missing translation for 'onlineSavingsMsg'
Learn More

MES, Monohydrate, Free Acidline, Crystal, Ultrapure Bioreagent, J.T. Baker™

Catalog No. p-5391218
Change view
Click to view available options
Quantity:
200 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003324 200 g Poly Bottle
02003326 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003324 Supplier Avantor Supplier No. 401402

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology Applications

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
Synonym mes monohydrate, 2-morpholinoethanesulfonic acid hydrate, mes hydrate, mes buffer hydrate, 4-morpholineethanesulfonic acid monohydrate, 4-morpholineethanesulfonic acid, monohydrate, 2-4-morpholinyl ethanesulfonic acid hydrate, mes hydrate buffer, 4-morpholineethanesulfonicacid, hydrate 1:1, mesmonohydrate
PubChem CID 16218417
IUPAC Name 2-(morpholin-4-yl)ethane-1-sulfonic acid hydrate
SMILES O.OS(=O)(=O)CCN1CCOCC1

Specifications

Packaging Poly Bottle
MDL Number MFCD00149409
Quantity 200 g
Formula Weight 213.25
Grade Ultrapure Bioreagent
Physical Form Crystals
Chemical Name or Material MES, Monohydrate, Free Acidline