missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MES, Monohydrate, Free Acidline, Crystal, Ultrapure Bioreagent, J.T. Baker™
Description
- For Liquid Chromatography and Molecular Biology Applications
Specifications
Specifications
| Packaging | Poly Bottle |
| MDL Number | MFCD00149409 |
| Quantity | 500 g |
| Formula Weight | 213.25 |
| Percent Purity | 100% |
| Grade | Ultrapure Bioreagent |
| Physical Form | Crystals |
| Chemical Name or Material | MES, Monohydrate, Free Acidline |