Learn More
Mercury(II) trifluoromethanesulfonate, 98%
CAS: 49540-00-3 | C2F6HgO6S2 | 498.718 g/mol
Supplier: Thermo Scientific Chemicals 03930506
Description
Hg(OTf)2 exhibits remarkable catalytic activity for the hydroxylative cyclization of 1,6-enynes. Mercuric triflate is a very versatile reagent and has been used for several organic catalytic transformations including C-C bond forming cyclizations, alkyne hydrations, heterocycle synthesis and very recently in C-N bond forming reactions. Commonly used as the metal source for Lewis acid catalyzed asymmetric reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Mercury(II) trifluoromethanesulfonate | |
| >350°C | |
| C2F6HgO6S2 | |
| MFCD00144746 | |
| UN2025 | |
| mercury ii trifluoromethanesulfonate, mercury triflate, mercury 2+ ditriflate, hg otf 2, ksc491a8p, mercury ii trifluoromethanesulphonate, mercury 2+ ; trifluoromethanesulfonate, mercury 2+ ion ditrifluoromethanesulfonate, mercury ii trifluoromethanesulfonate trace metals basis | |
| BPVYMDMPLCOQPJ-UHFFFAOYSA-L | |
| mercury(2+);trifluoromethanesulfonate | |
| 2775250 | |
| 98% |
| 49540-00-3 | |
| White | |
| Hg(CF3SO3)2 | |
| 5 g | |
| Moisture sensitive | |
| It is soluble in H2O and CH3CN,fairly soluble in CH3NO2 and CH2Cl2,insoluble in hexane,ether,and toluene. | |
| C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Hg+2] | |
| 498.718 | |
| 498.72 | |
| Powder |
Chemical Identifiers
| 49540-00-3 | |
| 498.718 | |
| BPVYMDMPLCOQPJ-UHFFFAOYSA-L | |
| 2775250 | |
| C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Hg+2] |
| C2F6HgO6S2 | |
| MFCD00144746 | |
| mercury ii trifluoromethanesulfonate, mercury triflate, mercury 2+ ditriflate, hg otf 2, ksc491a8p, mercury ii trifluoromethanesulphonate, mercury 2+ ; trifluoromethanesulfonate, mercury 2+ ion ditrifluoromethanesulfonate, mercury ii trifluoromethanesulfonate trace metals basis | |
| mercury(2+);trifluoromethanesulfonate |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
H300+H310+H330-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only