missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) trifluoromethanesulfonate, 98%

Catalog No. AA3930503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3930503 1 g
AA3930506 5 g
AA3930514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3930503 Supplier Thermo Scientific Chemicals Supplier No. 03930503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 49540-00-3 | C2F6HgO6S2 | 498.718 g/mol

Hg(OTf)2 exhibits remarkable catalytic activity for the hydroxylative cyclization of 1,6-enynes. Mercuric triflate is a very versatile reagent and has been used for several organic catalytic transformations including C-C bond forming cyclizations, alkyne hydrations, heterocycle synthesis and very recently in C-N bond forming reactions. Commonly used as the metal source for Lewis acid catalyzed asymmetric reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 49540-00-3
Molecular Formula C2F6HgO6S2
Molecular Weight (g/mol) 498.718
MDL Number MFCD00144746
InChI Key BPVYMDMPLCOQPJ-UHFFFAOYSA-L
Synonym mercury ii trifluoromethanesulfonate, mercury triflate, mercury 2+ ditriflate, hg otf 2, ksc491a8p, mercury ii trifluoromethanesulphonate, mercury 2+ ; trifluoromethanesulfonate, mercury 2+ ion ditrifluoromethanesulfonate, mercury ii trifluoromethanesulfonate trace metals basis
PubChem CID 2775250
IUPAC Name mercury(2+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Hg+2]

Specifications

Melting Point >350°C
Color White
Linear Formula Hg(CF3SO3)2
Quantity 1 g
UN Number UN2025
Sensitivity Moisture sensitive
Solubility Information It is soluble in H2O and CH3CN,fairly soluble in CH3NO2 and CH2Cl2,insoluble in hexane,ether,and toluene.
Formula Weight 498.72
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Mercury(II) trifluoromethanesulfonate
Show More Show Less
Hazard Category H300+H310+H330-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only