missing translation for 'onlineSavingsMsg'
Learn More

Mercuric Nitrate, 0.0141 N, 0.00705 M, 1 mL = 0.5 mg Cl-, Ricca Chemical

Supplier:  Ricca Chemical Company 47051

 View more versions of this product

Catalog No. 47051


Only null left
Explore available promotions

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.


Description

Description

Specifications

Specifications

Mercuric Nitrate
7783-34-8,7732-18-5,7697-37-2
0.2,100.0,0.35
MFCD00149736
KVICROHOONHSRH-UHFFFAOYSA-N
mercury(2+) hydrate dinitrate
1 mL = 0.5 mg Cl
Laboratory
4 L
0.0141Normal,0.00705Molar
Colorless
0.19,97.0,0.34
H2HgN2O7
mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
342.62
3084029
Amber Glass Bottle
Liquid

Chemical Identifiers

7783-34-8
342.62
KVICROHOONHSRH-UHFFFAOYSA-N
3084029
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
H2HgN2O7
MFCD00149736
mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
mercury(2+) hydrate dinitrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions