missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercuric Chloride, Saturated, Ricca Chemical
Supplier: Ricca Chemical Company R46700001C
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Specifications
| Mercuric Chloride | |
| Colorless | |
| 92.0,5.77 | |
| Liquid | |
| Cl2Hg | |
| mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2 | |
| [Cl-].[Cl-].[Hg++] | |
| 271.49 | |
| CHEBI:31823 | |
| Amber Glass Bottle |
| Saturated | |
| 7732-18-5,7487-94-7 | |
| 96.0,6.0 | |
| 1 L | |
| MFCD00011041 | |
| LWJROJCJINYWOX-UHFFFAOYSA-L | |
| mercury(2+) dichloride | |
| 24085 | |
| Laboratory |
Chemical Identifiers
| 7732-18-5 | |
| 271.49 | |
| LWJROJCJINYWOX-UHFFFAOYSA-L | |
| 24085 | |
| mercury(2+) dichloride |
| Cl2Hg | |
| MFCD00011041 | |
| mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2 | |
| CHEBI:31823 | |
| [Cl-].[Cl-].[Hg++] |
Safety and Handling
ShelfLife : 24 months