missing translation for 'onlineSavingsMsg'
Learn More

Mercaptoacetic acid, sodium salt, 98%

Catalog No. p-155688
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148890500 50 g Glass bottle
AC148892500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC148890500 Supplier Thermo Scientific Chemicals Supplier No. 148890500
Only null left

CAS: 367-51-1 | C2H3NaO2S | 114.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 367-51-1
Molecular Formula C2H3NaO2S
Molecular Weight (g/mol) 114.09
MDL Number MFCD00043386
InChI Key GNBVPFITFYNRCN-UHFFFAOYSA-M
Synonym sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
PubChem CID 23690444
ChEBI CHEBI:86481
IUPAC Name sodium;2-sulfanylacetate
SMILES C(C(=O)[O-])S.[Na+]

Specifications

Melting Point >300.0°C
Assay Percent Range 97.5% min. (Iodimetry)
Packaging Glass bottle
Linear Formula HSCH2CO2Na
Quantity 50 g
Merck Index 15,8819
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 114.09
Percent Purity 98%
Chemical Name or Material Mercaptoacetic acid, sodium salt, 98%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Absorb spillage to prevent material damage.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin
EINECSNumber 206-696-4
RTECSNumber AI7700000
TSCA TSCA

RUO – Research Use Only