Learn More
Mercaptoacetic acid sodium salt, 98%
CAS: 367-51-1 | C2H3NaO2S | 114.094 g/mol
Supplier: Thermo Scientific Chemicals J60290A1
Description
Reagent that protects tryptophan in amino acid analysis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Mercaptoacetic acid sodium salt | |
| >300°C | |
| C2H3NaO2S | |
| MFCD00043386 | |
| UN2811 | |
| Air sensitive; Hygroscopic | |
| GNBVPFITFYNRCN-UHFFFAOYSA-M | |
| sodium;2-sulfanylacetate | |
| 23690444 | |
| 114.1 | |
| Powder |
| 367-51-1 | |
| Glass bottle | |
| HSCH2CO2Na | |
| 1 kg | |
| 4569109 | |
| sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate | |
| C(C(=O)[O-])S.[Na+] | |
| 114.094 | |
| CHEBI:86481 | |
| 98% |
Chemical Identifiers
| 367-51-1 | |
| 114.094 | |
| GNBVPFITFYNRCN-UHFFFAOYSA-M | |
| 23690444 | |
| sodium;2-sulfanylacetate |
| C2H3NaO2S | |
| MFCD00043386 | |
| sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate | |
| CHEBI:86481 | |
| C(C(=O)[O-])S.[Na+] |
Safety and Handling
P234-P261-P264b-P270-P272-P280-P301+P310-P302+P352-P312-P330-P333+P313-P363-P390-P501c
H290-H301-H312-H317
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 206-696-4
RTECSNumber : AI7700000
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only