missing translation for 'onlineSavingsMsg'
Learn More

Mercaptoacetic acid sodium salt, 98%

Catalog No. AAJ6029014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Glass bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ6029014 25 g Plastic bottle
AAJ6029022 100 g Plastic bottle
AAJ6029036 500 g Plastic bottle
AAJ60290A1 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ6029014 Supplier Thermo Scientific Chemicals Supplier No. J6029014

CAS: 367-51-1 | C2H3NaO2S | 114.094 g/mol

Reagent that protects tryptophan in amino acid analysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 367-51-1
Molecular Formula C2H3NaO2S
Molecular Weight (g/mol) 114.094
MDL Number MFCD00043386
InChI Key GNBVPFITFYNRCN-UHFFFAOYSA-M
Synonym sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
PubChem CID 23690444
ChEBI CHEBI:86481
IUPAC Name sodium;2-sulfanylacetate
SMILES C(C(=O)[O-])S.[Na+]

Specifications

Melting Point >300°C
Packaging Plastic bottle
Linear Formula HSCH2CO2Na
Quantity 25 g
UN Number UN2811
Beilstein 4569109
Sensitivity Air sensitive; Hygroscopic
Formula Weight 114.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Mercaptoacetic acid sodium salt
Show More Show Less
Hazard Category H290-H301-H312-H317
Precautionary Statement P234-P261-P264b-P270-P272-P280-P301+P310-P302+P352-P312-P330-P333+P313-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 206-696-4
RTECSNumber AI7700000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only