missing translation for 'onlineSavingsMsg'
Learn More

Mequitazine 98.0+%, TCI America™

Catalog No. M2807100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2807100MG 100 mg
M28071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2807100MG Supplier TCI America Supplier No. M2807100MG

Chemical Identifiers

CAS 29216-28-2
Molecular Formula C20H22N2S
Molecular Weight (g/mol) 322.47
MDL Number MFCD01706743
InChI Key HOKDBMAJZXIPGC-UHFFFAOYSA-N
Synonym 10-(Quinuclidin-3-ylmethyl)-10H-phenothiazine, 10-(1-Azabicyclo[2.2.2]oct-3-ylmethyl)-10H-phenothiazine
PubChem CID 4066
ChEBI CHEBI:31821
IUPAC Name 10-(1-azabicyclo[2.2.2]octan-3-ylmethyl)phenothiazine
SMILES C1CN2CCC1C(C2)CN3C4=CC=CC=C4SC5=CC=CC=C53

Specifications

Melting Point 148°C
Color White
Quantity 100 mg
UN Number 2811
Formula Weight 322.47
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Mequitazine
RTECSNumber SP5080000
TSCA No