missing translation for 'onlineSavingsMsg'
Learn More

Mepivacaine hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018683
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018683 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018683 Supplier Merck Emd Millipore Supplier No. PHR1955500MG
Only null left
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Mepivacaine hydrochloride is a local anesthetic that is related chemically and pharmacologically to the amide-type local anesthetics. It is often administered via local infiltration, peripheral nerve block, etc.

Specifications

CAS 1722-62-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C15H22N2 O · HCl
MDL Number MFCD00144738
Synonym Mepivacaine hydrochloride; 1-Methyl-2′,6′-pipecoloxylidine hydrochloride; N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide hydrochloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 282.81
Grade Certified Reference Material