missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Menthyl Chloroformate 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 109°C |
| Quantity | 5 mL |
| UN Number | 3277 |
| Formula Weight | 218.72 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | (-)-Menthyl Chloroformate |
Safety and Handling
| TSCA | No |