accessibility menu, dialog, popup

UserName

CAS RN 14602-86-9

H3CCH3CH3OClO

CAS RN 14602-86-9

IUPAC Name: 5-methyl-2-(propan-2-yl)cyclohexyl carbonochloridate
Synonyms: 2-isopropyl-5-methylcyclohexyl carbonochloridate
Molecular Weight (g/mol): 218.72
Molecular Formula: C11H19ClO2
InChi Key: KIUPCUCGVCGPPA-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C)CC1OC(Cl)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results