missing translation for 'onlineSavingsMsg'
Learn More

(+)-Menthyl Acetate 98.0+%, TCI America™

Catalog No. M26311ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26311ML 1 mL
M26315ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26311ML Supplier TCI America Supplier No. M26311ML

Chemical Identifiers

CAS 5157-89-1
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00062977
InChI Key XHXUANMFYXWVNG-DLOVCJGASA-N
Synonym neoisomenthyl acetate, +-menthylacetate, 1s-2alpha-1-methylethyl-5alpha-methylcyclohexan-1alpha-ol acetate
PubChem CID 21630934
IUPAC Name [(1S,2S,5S)-5-methyl-2-propan-2-ylcyclohexyl] acetate
SMILES CC1CCC(C(C1)OC(=O)C)C(C)C

Specifications

Color Colorless
Boiling Point 229°C
Quantity 1 mL
Formula Weight 198.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (+)-Menthyl Acetate