missing translation for 'onlineSavingsMsg'
Learn More

(+)-Menthone, ≥98.5% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111013964
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013964 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013964 Supplier Merck Emd Millipore Supplier No. 636751MLF
Only null left
Featured Industries: Food and Beverages

Specifications

CAS 3391-87-5
Density 0.893 g/mL (at 20°C)
Flash Point 72°C - closed cup
Physical Form Neat
Linear Formula C10H18 O
MDL Number MFCD00210063
Boiling Point 209°C to 210°C
Synonym (1 S,4 R)-p-Menthan-3-one; (2 R,5 S)-2-Isopropyl-5-methylcyclohexanone
Type Analytical Standard
Quantity 1 mL
Formula Weight 154.25
Percent Purity ≥98.5% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less