missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-Menthol 98.0+%, TCI America™
Supplier: TCI America M0321100ML
| Quantity | 100 mL |
|---|
Chemical Identifiers
| C10H20O | |
| MFCD00001484 | |
| menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol | |
| (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol |
Specifications
| (+/-)-Menthol | |
| White | |
| 100 mL | |
| menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol | |
| CC1CCC(C(C1)O)C(C)C | |
| 156.269 | |
| 156.27 | |
| Crystals |
| 89-78-1 | |
| C10H20O | |
| MFCD00001484 | |
| NOOLISFMXDJSKH-IVZWLZJFSA-N | |
| (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol | |
| 6566020 | |
| ≥98.0% (GC) |
Safety and Handling
EINECSNumber : (3)-2333
RTECSNumber : OT0525000
TSCA : Yes