missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Menthol 98.0+%, TCI America™

Catalog No. p-7132099
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M032125ML 25 mL
M0321100ML 100 mL
M0321500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M032125ML Supplier TCI America Supplier No. M032125ML
Only null left

Chemical Identifiers

CAS 89-78-1
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00001484
InChI Key NOOLISFMXDJSKH-IVZWLZJFSA-N
Synonym menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
PubChem CID 6566020
IUPAC Name (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)C

Specifications

Color White
Quantity 25 mL
Formula Weight 156.27
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material (+/-)-Menthol
EINECSNumber (3)-2333
RTECSNumber OT0525000
TSCA Yes