missing translation for 'onlineSavingsMsg'
Learn More

(-)-Menthol, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. p-7187512
Change view
Click to view available options
Quantity:
1 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013566 1 g
111013567 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013566 Supplier Merck Emd Millipore Supplier No. 636601G
Only null left

(-)-Menthol is a cyclic terpene alcohol that occurs in mentha species.It is responsible for their distinctive odor and flavor.

Featured Industries: Food and Beverages

Specifications

CAS 2216-51-5
Flash Point 101°C - closed cup
Physical Form Neat
Linear Formula C10H20 O
MDL Number MFCD00062979
Melting Point 43°C
Synonym (1 R,2 S,5 R)-2-Isopropyl-5-methylcyclohexanol; 5-Methyl-2-(1-methylethyl)cyclohexanol
Type Analytical Standard
Quantity 1 g
Formula Weight 156.27
Percent Purity ≥99% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less