missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Meningococcus, Gram Negative

Catalog No. S30205
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30205 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30205 Supplier Eisco™ Supplier No. BS18094

Prepared microscope slide of gram-negative Meningococcus. Meningococcus is a bacteria causing meningitis in humans. Ideal addition to a microbacterial microscope slide collection.

  • Pre-cleaned glass slide with ground edges
  • Professionally stained samples for best visualization
  • Ideal for for biology classrooms to explore structure-function relationships as per NGSS standards
  • Shows characteristic morphology

Specifications

Biosafety Level 1
Concentration 10%
Color Clear
Material Glass
Mounting Type Mounting Medium
Type Microbiology slide
For Use With (Application) Microscopy
Manufacturing Origin China
Media Type Formalin
Packaging Cardboard Mailer
Quantity 1
Species Meningococcus
Size 1 x 3 in.
Slide Type Prepared Microbiology Slide
Show More Show Less