missing translation for 'onlineSavingsMsg'
Learn More

Menadione Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014749
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014749 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014749 Supplier Merck Emd Millipore Supplier No. PHR1411500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Menadione is an antihemorrhagic compound and it finds application in blood clotting and bone metabolism as an essential cofactor. Additionally, at relatively higher dose, menadione exhibits antitumor activity on several human cancer cells.

Specifications

CAS 58-27-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C11H8O2
MDL Number MFCD00001681
Melting Point 105°C to 107°C
Synonym Menadione; 2-Methyl-1,4-naphthoquinone; Vitamin K3
Type Certified Reference Material
Quantity 500 mg
Formula Weight 172.18
Grade Certified Reference Material
Show More Show Less