missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Stretch Denim Flame Resistant Dungaree

Men's Stretch Denim Flame Resistant Dungaree fits like your weekend jeans and moves with you while you work. Relaxed through waist, hip, and thigh to allow for easy movement.

Supplier:  Bulwark™ PSJ6SD3334

 View more versions of this product

Catalog No. 17-204-371

Only null left
Explore available promotions

Description

Description

  • Increased durability with triple needle felled out seams and seat seam
  • Sits below natural waist with relaxed fit through waist, hip, and thigh
  • Bootcut leg opening fits easily over boots
  • Five traditional jean style pockets, with an additional ruler pocket on right leg and utility pocket on left leg for extra storage
  • One-piece waistband with metal button closure (metal is not exposed to skin)
  • Utility hammer loop on left hip
  • Reinforced pockets help prevent holes
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 7
  • Primary Closure: Concealed Button
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 12 oz. (405 g/m2) Flame Resistant Denim
  • Blend: Body: 94% Cotton/5% Polyester/1% Spandex Elastane
  • Country of Origin: Imported, Made in USA Fabric
Specifications

Specifications

33 x 34 in.
Cotton, Polyester, Spandex
Stitched
Concealed Button
Category 2
NFPA 2112, NFPA 70E
Men's
Denim
6
Reusable
15
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions