missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Stretch Denim Flame Resistant Dungaree

Catalog No. p-200000038
Change view
Click to view available options
Size:
30 x 30 in.
30 x 32 in.
30 x 34 in.
32 x 30 in.
32 x 32 in.
32 x 34 in.
32 x 36 in.
33 x 30 in.
33 x 32 in.
33 x 34 in.
33 x 36 in.
34 x 30 in.
34 x 32 in.
34 x 34 in.
34 x 36 in.
35 x 30 in.
35 x 32 in.
35 x 34 in.
35 x 36 in.
36 x 30 in.
36 x 32 in.
36 x 34 in.
36 x 36 in.
38 x 30 in.
38 x 32 in.
38 x 34 in.
38 x 36 in.
40 x 30 in.
40 x 32 in.
40 x 34 in.
40 x 36 in.
42 x 30 in.
42 x 32 in.
42 x 34 in.
42 x 36 in.
44 x 30 in.
44 x 32 in.
44 x 34 in.
44 x 36 in.
46 x 30 in.
46 x 32 in.
46 x 34 in.
48 x 30 in.
48 x 32 in.
50 x 30 in.
50 x 32 in.
46 product options available for selection
Product selection table with 46 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204233 30 x 30 in.
17203943 30 x 32 in.
17204032 30 x 34 in.
17204222 32 x 30 in.
17204224 32 x 32 in.
17204050 32 x 34 in.
17204165 32 x 36 in.
17204363 33 x 30 in.
17204453 33 x 32 in.
17204371 33 x 34 in.
17204372 33 x 36 in.
17204230 34 x 30 in.
17204020 34 x 32 in.
17204215 34 x 34 in.
17204121 34 x 36 in.
17204446 35 x 30 in.
17204249 35 x 32 in.
17204448 35 x 34 in.
17204132 35 x 36 in.
17204449 36 x 30 in.
17204350 36 x 32 in.
17204258 36 x 34 in.
17204266 36 x 36 in.
17204314 38 x 30 in.
17203972 38 x 32 in.
17204023 38 x 34 in.
17204478 38 x 36 in.
17203961 40 x 30 in.
17203963 40 x 32 in.
17203978 40 x 34 in.
17204316 40 x 36 in.
17203985 42 x 30 in.
17204475 42 x 32 in.
17204257 42 x 34 in.
17203974 42 x 36 in.
17204275 44 x 30 in.
17204309 44 x 32 in.
17204262 44 x 34 in.
17203964 44 x 36 in.
17204174 46 x 30 in.
17204279 46 x 32 in.
17204473 46 x 34 in.
17203988 48 x 30 in.
17204271 48 x 32 in.
17204307 50 x 30 in.
17204467 50 x 32 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 46 options available.
46 options
Catalog No. 17204233 Supplier Bulwark™ Supplier No. PSJ6SD 30 30
Only null left

Men's Stretch Denim Flame Resistant Dungaree fits like your weekend jeans and moves with you while you work. Relaxed through waist, hip, and thigh to allow for easy movement.

  • Increased durability with triple needle felled out seams and seat seam
  • Sits below natural waist with relaxed fit through waist, hip, and thigh
  • Bootcut leg opening fits easily over boots
  • Five traditional jean style pockets, with an additional ruler pocket on right leg and utility pocket on left leg for extra storage
  • One-piece waistband with metal button closure (metal is not exposed to skin)
  • Utility hammer loop on left hip
  • Reinforced pockets help prevent holes
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 7
  • Primary Closure: Concealed Button
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 12 oz. (405 g/m2) Flame Resistant Denim
  • Blend: Body: 94% Cotton/5% Polyester/1% Spandex Elastane
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size 30 x 30 in.
Gender Men's
Material Cotton, Polyester, Spandex
Color Denim
Seam Type Stitched
No. of Pockets 6
Closure Type Concealed Button
Disposable Reusable
Hazard Risk Category Category 2
Arc Flash Rating 15
Certifications/Compliance NFPA 2112, NFPA 70E
Show More Show Less