missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Straight Fit Bootcut Flame Resistant Jean with Stretch
Men's Straight Fit Flame Resistant Jean with Stretch are made from 12 oz. flame-resistant 94% cotton, 5% polyester, 1% spandex denim that provides comfort and mobility while you work.
Supplier: Bulwark™ PSJ4SD3432
Description
- Straight fit with a larger waistband for comfort and durability
- One-piece waistband with concealed, metal button closure
- Two patch spade shaped hip pockets and scoop front pockets
- Larger belt loop and waistband
- Stretch fabric with felled inseam and seat seam
- Hazard: Flash Fire, Arc Flash
- Wash Care: Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame-resistant, 12 oz. (405 g/m2)
- Blend: 94% Cotton/5% Polyester/1% Spandex
- Country of Origin: Imported, Made in USA Fabric
Specifications
| 34 x 32 in. | |
| Cotton, Polyester, Spandex | |
| Stitched | |
| Reusable | |
| 15 |
| Men's | |
| Denim | |
| Button | |
| Category 2 | |
| NFPA 2112 |