missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Personal Protective Equipment
- Safety Clothing
- Pants, Trousers, and Leggings
- Heat and Fire Resistant Pants, Trousers, and Leggings
- Bulwark™ Men's Straight Fit Bootcut Flame Resistant Jean with Stretch
Bulwark™ Men's Straight Fit Bootcut Flame Resistant Jean with Stretch
Catalog No. p-200000036
Change view
Click to view available options
Size:
30 x 30 in.
30 x 32 in.
30 x 34 in.
32 x 30 in.
32 x 32 in.
32 x 34 in.
32 x 36 in.
33 x 30 in.
33 x 32 in.
33 x 34 in.
33 x 36 in.
34 x 30 in.
34 x 32 in.
34 x 34 in.
34 x 36 in.
35 x 30 in.
35 x 34 in.
35 x 36 in.
36 x 32 in.
36 x 34 in.
36 x 36 in.
38 x 30 in.
38 x 32 in.
38 x 34 in.
38 x 36 in.
40 x 30 in.
40 x 32 in.
40 x 34 in.
40 x 36 in.
42 x 30 in.
42 x 32 in.
42 x 34 in.
42 x 36 in.
44 x 30 in.
44 x 32 in.
44 x 34 in.
44 x 36 in.
46 x 30 in.
46 x 32 in.
46 x 34 in.
48 x 32 in.
48 x 34 in.
50 x 30 in.
50 x 32 in.
50 x 34 in.
45 product options available for selection
| Catalog No. | Size |
|---|---|
| 17204187 | 30 x 30 in. |
| 17204170 | 30 x 32 in. |
| 17204407 | 30 x 34 in. |
| 17204469 | 32 x 30 in. |
| 17204472 | 32 x 32 in. |
| 17204379 | 32 x 34 in. |
| 17203973 | 32 x 36 in. |
| 17204261 | 33 x 30 in. |
| 17204403 | 33 x 32 in. |
| 17204313 | 33 x 34 in. |
| 17204268 | 33 x 36 in. |
| 17204375 | 34 x 30 in. |
| 17204324 | 34 x 32 in. |
| 17204398 | 34 x 34 in. |
| 17204308 | 34 x 36 in. |
| 17204406 | 35 x 30 in. |
| 17204411 | 35 x 34 in. |
| 17204474 | 35 x 36 in. |
| 17204185 | 36 x 32 in. |
| 17204168 | 36 x 34 in. |
| 17203967 | 36 x 36 in. |
| 17204323 | 38 x 30 in. |
| 17204300 | 38 x 32 in. |
| 17204206 | 38 x 34 in. |
| 17203991 | 38 x 36 in. |
| 17204433 | 40 x 30 in. |
| 17204331 | 40 x 32 in. |
| 17204388 | 40 x 34 in. |
| 17204007 | 40 x 36 in. |
| 17204114 | 42 x 30 in. |
| 17204236 | 42 x 32 in. |
| 17204328 | 42 x 34 in. |
| 17204107 | 42 x 36 in. |
| 17204332 | 44 x 30 in. |
| 17204109 | 44 x 32 in. |
| 17204092 | 44 x 34 in. |
| 17204115 | 44 x 36 in. |
| 17204073 | 46 x 30 in. |
| 17204397 | 46 x 32 in. |
| 17204214 | 46 x 34 in. |
| 17204451 | 48 x 32 in. |
| 17204349 | 48 x 34 in. |
| 17204067 | 50 x 30 in. |
| 17204031 | 50 x 32 in. |
| 17204394 | 50 x 34 in. |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 45 options available.
Description
- Straight fit with a larger waistband for comfort and durability
- One-piece waistband with concealed, metal button closure
- Two patch spade shaped hip pockets and scoop front pockets
- Larger belt loop and waistband
- Stretch fabric with felled inseam and seat seam
- Hazard: Flash Fire, Arc Flash
- Wash Care: Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame-resistant, 12 oz. (405 g/m2)
- Blend: 94% Cotton/5% Polyester/1% Spandex
- Country of Origin: Imported, Made in USA Fabric
Specifications
Specifications
| Size | 30 x 30 in. |
| Gender | Men's |
| Material | Cotton, Polyester, Spandex |
| Color | Denim |
| Seam Type | Stitched |
| Closure Type | Button |
| Disposable | Reusable |
| Hazard Risk Category | Category 2 |
| Arc Flash Rating | 15 |
| Certifications/Compliance | NFPA 2112 |