missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Relaxed Fit Bootcut Flame Resistant Jean with Stretch

Men's Relaxed Fit Bootcut Flame Resistant Jean with Stretch are made from 12 oz. flame-resistant 94% cotton, 5% polyester, 1% spandex denim that provides comfort and mobility while you work.

Supplier:  Bulwark™ PSJ2SD4430

 View more versions of this product

Catalog No. 17-204-413

Only null left
Explore available promotions

Description

Description

  • Sits below natural waist with relaxed fit through waist, hip and thigh
  • Bootcut leg opening fits easily over boots
  • One-piece waistband with metal button closure (metal is not exposed to skin)
  • Two hip pockets and scoop front pockets
  • Larger belt loop and waistband
  • Contrasting, two-toned stitching
  • Stretch fabric inseam and seat seam
  • Hazard: Flash Fire, Arc Flash
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame-resistant, 12 oz. (405 g/m2)
  • Blend: 94% Cotton/5% Polyester/1% Spandex
  • Country of Origin: Imported, Made in USA Fabric
Specifications

Specifications

44 x 30 in.
Cotton, Polyester, Spandex
Stitched
Reusable
15
Men's
Denim
Zipper
Category 2
NFPA 2112
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions