missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Personal Protective Equipment
- Safety Clothing
- Pants, Trousers, and Leggings
- Heat and Fire Resistant Pants, Trousers, and Leggings
- Bulwark™ Men's Relaxed Fit Bootcut Flame Resistant Jean with Stretch
Bulwark™ Men's Relaxed Fit Bootcut Flame Resistant Jean with Stretch
Catalog No. p-200000034
Change view
Click to view available options
Size:
30 x 30 in.
30 x 32 in.
30 x 34 in.
32 x 30 in.
32 x 32 in.
32 x 34 in.
32 x 36 in.
33 x 30 in.
33 x 32 in.
33 x 34 in.
33 x 36 in.
34 x 30 in.
34 x 32 in.
34 x 34 in.
34 x 36 in.
35 x 30 in.
35 x 32 in.
35 x 34 in.
35 x 36 in.
36 x 30 in.
36 x 32 in.
36 x 34 in.
36 x 36 in.
38 x 30 in.
38 x 32 in.
38 x 34 in.
38 x 36 in.
40 x 30 in.
40 x 32 in.
40 x 34 in.
42 x 30 in.
42 x 32 in.
42 x 36 in.
44 x 30 in.
44 x 32 in.
44 x 34 in.
44 x 36 in.
46 x 30 in.
46 x 32 in.
46 x 34 in.
48 x 30 in.
48 x 32 in.
48 x 34 in.
50 x 30 in.
50 x 32 in.
50 x 34 in.
47 product options available for selection
| Catalog No. | Size |
|---|---|
| 17204034 | 30 x 30 in. |
| 17204075 | 30 x 32 in. |
| 17204035 | 30 x 34 in. |
| 17204286 | 32 x 30 in. |
| 17204219 | 32 x 32 in. |
| 17204293 | 32 x 34 in. |
| 17204157 | 32 x 36 in. |
| 17204076 | 33 x 30 in. |
| 17204079 | 33 x 32 in. |
| 17204285 | 33 x 34 in. |
| 17204364 | 33 x 36 in. |
| 17204220 | 34 x 30 in. |
| 17204151 | 34 x 32 in. |
| 17204229 | 34 x 34 in. |
| 17204052 | 34 x 36 in. |
| 17204345 | 35 x 30 in. |
| 17204145 | 35 x 32 in. |
| 17204065 | 35 x 34 in. |
| 17204155 | 35 x 36 in. |
| 17204140 | 36 x 30 in. |
| 17204078 | 36 x 32 in. |
| 17204361 | 36 x 34 in. |
| 17204144 | 36 x 36 in. |
| 17204217 | 38 x 30 in. |
| 17204352 | 38 x 32 in. |
| 17204070 | 38 x 34 in. |
| 17204072 | 38 x 36 in. |
| 17204164 | 40 x 30 in. |
| 17204283 | 40 x 32 in. |
| 17204146 | 40 x 34 in. |
| 17204289 | - |
| 17204297 | 42 x 30 in. |
| 17204169 | 42 x 32 in. |
| 17204317 | 42 x 36 in. |
| 17204413 | 44 x 30 in. |
| 17204254 | 44 x 32 in. |
| 17204461 | 44 x 34 in. |
| 17204464 | 44 x 36 in. |
| 17204402 | 46 x 30 in. |
| 17204277 | 46 x 32 in. |
| 17204175 | 46 x 34 in. |
| 17204022 | 48 x 30 in. |
| 17204476 | 48 x 32 in. |
| 17204298 | 48 x 34 in. |
| 17204260 | 50 x 30 in. |
| 17204299 | 50 x 34 in. |
| 17204189 | 50 x 32 in. |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 47 options available.
Description
- Sits below natural waist with relaxed fit through waist, hip and thigh
- Bootcut leg opening fits easily over boots
- One-piece waistband with metal button closure (metal is not exposed to skin)
- Two hip pockets and scoop front pockets
- Larger belt loop and waistband
- Contrasting, two-toned stitching
- Stretch fabric inseam and seat seam
- Hazard: Flash Fire, Arc Flash
- Wash Care: Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame-resistant, 12 oz. (405 g/m2)
- Blend: 94% Cotton/5% Polyester/1% Spandex
- Country of Origin: Imported, Made in USA Fabric