missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Navy Flame Resistant Flex Knit Henley
Men's Navy Flame Resistant Flex Knit Henley has a casual, contemporary look with lightweight fabric for added comfort and range of movement, curved flatlock seams for optimal fit, and a unique front pocket design with pencil stall.
$131.29 - $131.29
Specifications
| Gender | Men's |
|---|---|
| Material | Cotton, Spandex |
| Color | Navy |
| Neck Style | Henley |
| Sleeve Style | Long Sleeve |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-204-235
|
Bulwark™
STL2NVRGS |
Small |
Each for $131.29
|
|
|||||
|
17-204-391
|
Bulwark™
STL2NVRGM |
Medium |
Each for $131.29
|
|
|||||
|
17-204-090
|
Bulwark™
STL2NVRGL |
Large |
Each for $131.29
|
|
|||||
|
17-204-392
|
Bulwark™
STL2NVRGXL |
X-Large |
Each for $131.29
|
|
|||||
|
17-204-242
|
Bulwark™
STL2NVRGXXL |
2X-Large |
Each for $131.29
|
|
|||||
|
17-204-342
|
Bulwark™
STL2NVLNL |
Large (Long) |
Each for $131.29
|
|
|||||
|
17-103-742
|
Bulwark™
STL2NVLNXL |
X-Large (Long) |
Each for $131.29
|
|
|||||
|
17-103-741
|
Bulwark™
STL2NVLNXXL |
2X-Large (Long) |
Each for $131.29
|
|
|||||
Description
- Three-button front placket for enhanced appearance
- Contrasting pattern on sleeve for athletic look
- CAT 2/2112 left sleeve label leaves no question that you are protected on the job site
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: Body: 6.75 oz. (230 g/m2) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m2) Flame Resistant Jersey
- Blend: Body: 93% Cotton/7% Spandex Elastane, Contrast Panels: 93% Cotton/7% Spandex Elastane
- Collar: Rib
- Country of Origin: Imported, Made in USA Fabric
Specifications
| Men's | |
| Navy | |
| Long Sleeve | |
| Button | |
| 1 | |
| 8.3 |
| Cotton, Spandex | |
| Henley | |
| Knit | |
| Reusable | |
| Category 2 | |
| NFPA 2112, NFPA 70E |