missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Navy Flame Resistant Flex Knit Henley

Catalog No. 17204235
Change view
Click to view available options
Size:
2X-Large (Long)
Large (Long)
X-Large (Long)
Small
Medium
Large
X-Large
2X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204235 Small
17204391 Medium
17204090 Large
17204392 X-Large
17204242 2X-Large
17204342 Large (Long)
17103742 X-Large (Long)
17103741 2X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17204235 Supplier Bulwark™ Supplier No. STL2NV RG S

Men's Navy Flame Resistant Flex Knit Henley has a casual, contemporary look with lightweight fabric for added comfort and range of movement, curved flatlock seams for optimal fit, and a unique front pocket design with pencil stall.

  • Three-button front placket for enhanced appearance
  • Contrasting pattern on sleeve for athletic look
  • CAT 2/2112 left sleeve label leaves no question that you are protected on the job site
  • Hazard: Flash Fire, Arc Flash
  • Wash Care: Home Wash
  • Fabric: Body: 6.75 oz. (230 g/m2) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m2) Flame Resistant Jersey
  • Blend: Body: 93% Cotton/7% Spandex Elastane, Contrast Panels: 93% Cotton/7% Spandex Elastane
  • Collar: Rib
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size Small
Gender Men's
Material Cotton, Spandex
Color Navy
Neck Style Henley
Sleeve Style Long Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 1
Hazard Risk Category Category 2
Arc Flash Rating 8.3
Certifications/Compliance NFPA 2112, NFPA 70E
Show More Show Less