missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Lightweight Excel FR™ ComforTouch™ Royal Premium Flame Resistant Coverall

Men's Lightweight Excel FR™ ComforTouch™ Royal Premium Flame Resistant Coverall has eight total pockets to keep your tools at hand, while elastic waist inserts and deep-pleated action back provide added mobility and comfort while you work.

Supplier:  Bulwark™ CLB2RBRG3XL

 View more versions of this product

Catalog No. 17-204-209

Only null left
Explore available promotions

Description

Description

  • This lightweight premium coverall provides CAT 2 FR/AR protection without added bulk.
  • Innovative features provide extra comfort and convenience.
  • Eight total pockets keep your tools at hand, while elastic waist inserts and deep-pleated action back provide added mobility and comfort while you work.
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Primary Closure: Concealed Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 7 oz. (235 g/m&superscript_2;)
  • Blend: 88% Cotton/ 12% Nylon
  • Closure: Two-way concealed Nomex™ taped brass break-away zipper, concealed snap at top of zipper and at neck
  • Pocket: Two front swing pockets are topstitched and lined and two patch hip pockets have single concealed snap closure, One left sleeve pocket, two patch chest pockets have flaps and concealed snap closure and a rule pocket on right leg
  • Features: Two-way concealed Nomex™ taped brass break-away zipper, concealed snap at top of zipper and at neck, Two-piece, lay-flay collar, deep-pleated action back and elastic waist inserts, Hemmed sleeve with adjustable snap and side vent opening with concealed snap closure
  • Product Collection: Excel FR Comfortouch
  • Protection: ATPV Arc Rating: 8.6
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Designed to be worn over other clothing. Fit over work boots.
  • Gender: Male
  • Materials: Cotton/Nylon
  • Brand: Bulwark™
Specifications

Specifications

Flame-resistant Coveralls
Men's
Royal Blue
Elastic (Stitched-in)
Open
Unhooded
Stitched
CAT 2
8.6
8
3X-Large
Cotton, Nylon
Reusable
Open
Collared
2-way Front Zipper with Storm Flap
Non-sterile
Flame
NFPA 2112
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions