missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Lightweight Excel FR™ ComforTouch™ Royal Premium Flame Resistant Coverall

Catalog No. p-200000024
Change view
missing translation for 'orderingAttributeHoverText'
Size:
2X-Large
2X-Large (Long)
3X-Large
3X-Large (Long)
Large
Large (Long)
Medium
X-Large
X-Large (Long)
Catalog No. Size
17204444 Medium
17204101 Large
17204443 X-Large
17204240 2X-Large
17204209 3X-Large
17204241 Large (Long)
17204119 X-Large (Long)
17204442 2X-Large (Long)
17204208 3X-Large (Long)
9 options

Catalog No. 17204444

Supplier: Bulwark™ CLB2RB RG M

Only null left

Men's Lightweight Excel FR™ ComforTouch™ Royal Premium Flame Resistant Coverall has eight total pockets to keep your tools at hand, while elastic waist inserts and deep-pleated action back provide added mobility and comfort while you work.

  • This lightweight premium coverall provides CAT 2 FR/AR protection without added bulk.
  • Innovative features provide extra comfort and convenience.
  • Eight total pockets keep your tools at hand, while elastic waist inserts and deep-pleated action back provide added mobility and comfort while you work.
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Primary Closure: Concealed Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 7 oz. (235 g/m&superscript_2;)
  • Blend: 88% Cotton/ 12% Nylon
  • Closure: Two-way concealed Nomex™ taped brass break-away zipper, concealed snap at top of zipper and at neck
  • Pocket: Two front swing pockets are topstitched and lined and two patch hip pockets have single concealed snap closure, One left sleeve pocket, two patch chest pockets have flaps and concealed snap closure and a rule pocket on right leg
  • Features: Two-way concealed Nomex™ taped brass break-away zipper, concealed snap at top of zipper and at neck, Two-piece, lay-flay collar, deep-pleated action back and elastic waist inserts, Hemmed sleeve with adjustable snap and side vent opening with concealed snap closure
  • Product Collection: Excel FR Comfortouch
  • Protection: ATPV Arc Rating: 8.6
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Designed to be worn over other clothing. Fit over work boots.
  • Gender: Male
  • Materials: Cotton/Nylon
  • Brand: Bulwark™

Specifications

Product Type Flame-resistant Coveralls
Size Medium
Gender Men's
Material Cotton, Nylon
Color Royal Blue
Disposable Reusable
Waist Style Elastic (Stitched-in)
Ankle Style Open
Cuff Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Protection Class CAT 2
Resistance Flame
Arc Flash Rating 8.6
Certifications/Compliance NFPA 2112
No. of Pockets 8
Show More Show Less