missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Lightweight Excel FR™ ComfortTouch™ Khaki Premium Flame Resistant Coverall
These Khaki Premium Flame Resistant Coveralls have eight total pockets to keep your tools at hand, while elastic waist inserts and a deep-pleated action back provide added mobility and comfort while you work.
Supplier: Bulwark™ CLB2KHLNL
Description
- Designed to be worn over clothing
- Two front swing pockets are topstitched and lined and two patch hip pockets have single concealed snap closure
- One left sleeve pocket, two patch chest pockets have flaps and concealed snap closure and a rule pocket on right leg
- Two-way concealed Nomex™ taped brass break-away zipper, concealed snap at top of zipper and at neck
- Hemmed sleeve with adjustable snap and side vent opening with concealed snap closure
- Fits over work boots
- Hazard: Flash Fire, Arc Flash
- Primary Closure: Concealed Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame resistant, 7 oz. (235 g/m2)
- Blend: 88% Cotton/12% Nylon
- Country of Origin: Imported, Made in USA Fabric
Specifications
| Flame-resistant Coveralls | |
| Men's | |
| Khaki | |
| Elastic (Stitched-in) | |
| Open | |
| Unhooded | |
| Stitched | |
| CAT 2 | |
| 8.6 | |
| 8 |
| Large (Long) | |
| Cotton, Nylon | |
| Reusable | |
| Open | |
| Collared | |
| 2-way Front Zipper with Storm Flap | |
| Non-sterile | |
| Flame | |
| NFPA 2112 |