missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Lightweight Excel FR™ ComfortTouch™ Khaki Premium Flame Resistant Coverall

Catalog No. p-200000020
Change view
Click to view available options
Size:
2X-Large
2X-Large (Long)
Large
Large (Long)
Medium
X-Large
X-Large (Long)
Catalog No. Size
17103764 Medium
17204213 Large
17103763 X-Large
17204014 2X-Large
17204103 Large (Long)
17204337 X-Large (Long)
17204212 2X-Large (Long)
7 options

Catalog No. 17103764

Supplier: Bulwark™ CLB2KH RG M

Only null left

These Khaki Premium Flame Resistant Coveralls have eight total pockets to keep your tools at hand, while elastic waist inserts and a deep-pleated action back provide added mobility and comfort while you work.

  • Designed to be worn over clothing
  • Two front swing pockets are topstitched and lined and two patch hip pockets have single concealed snap closure
  • One left sleeve pocket, two patch chest pockets have flaps and concealed snap closure and a rule pocket on right leg
  • Two-way concealed Nomex™ taped brass break-away zipper, concealed snap at top of zipper and at neck
  • Hemmed sleeve with adjustable snap and side vent opening with concealed snap closure
  • Fits over work boots
  • Hazard: Flash Fire, Arc Flash
  • Primary Closure: Concealed Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 7 oz. (235 g/m2)
  • Blend: 88% Cotton/12% Nylon
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Product Type Flame-resistant Coveralls
Size Medium
Gender Men's
Material Cotton, Nylon
Color Khaki
Disposable Reusable
Waist Style Elastic (Stitched-in)
Ankle Style Open
Cuff Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Protection Class CAT 2
Resistance Flame
Arc Flash Rating 8.6
Certifications/Compliance NFPA 2112
No. of Pockets 8
Show More Show Less