missing translation for 'onlineSavingsMsg'
Learn More

Memantine Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014158
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014158 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014158 Supplier Merck Emd Millipore Supplier No. PHR18861G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Memantine Hydrochloride is a drug used for treating the Alzheimer′s disease by means of reducing abnormal activity in the brain. It can help patients suffering from dementia as it assists them to think more clearly and perform their chores with ease.

Specifications

CAS 41100-52-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C12H21 N · HCl
MDL Number MFCD00214336
Synonym Memantine hydrochloride; 3,5-Dimethyl-1-adamantanamine hydrochloride; 3,5-Dimethylamantadine hydrochloride; 3,5-Dimethyltricyclo[3.3.1.13,7 ]decan-1-amine hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 215.76
Grade Certified Reference Material