missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM Non-Essential Amino Acids Solution (100X)
GREENER_CHOICE

Catalog No. p-9807003
Encompass
Change view
Click to view available options
Quantity:
100 mL
20 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11140050 100 mL
11140076 20 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11140050 Supplier Gibco™ Supplier No. 11140050
Only null left

Gibco™ MEM Non-Essential Amino Acids are used as a supplement for cell culture medium, to increase cell growth and viability.

Gibco™ MEM Non-Essential Amino Acids are used as a supplement for cell culture medium, to increase cell growth and viability. Gibco™ MEM Non-Essential Amino Acids contains the same non-essential amino acids found in the standard Minimum Essential Medium (MEM) at a strength of 100X. The complete formulation is available.

Specifications

Content And Storage Store at 2–8°C. Protect from light.
Cell Type Mammalian Cells
Concentration 100X
Form Liquid
Product Type Amino Acid Supplement
Sterility Sterile-filtered
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
pH 0.9
Quantity 100 mL
Shipping Condition Room Temperature
Where can I find the formulation of MEM Non-Essential Amino Acids Solution (100X) (Cat. No. 11140050)?

The formulation of MEM Non-Essential Amino Acids Solution (100X) can be found at this link: https://www.thermofisher.com/us/en/home/technical-resources/media-formulation.165.html

What is the solvent used for MEM Non-Essential Amino Acids Solution (100X)?

The solvent is distilled water.

What is Gibco MEM Non-Essential Amino Acids?

MEM Non-Essential Amino Acids are used as a supplement for cell culture medium, to increase cell growth and viability. MEM Non-Essential Amino Acids contains the same non-essential amino acids found in the standard Minimum Essential Medium (MEM) at a strength of 100X. The complete formulation is available here (http://www.thermofisher.com/us/en/home/technical-resources/media-formulation.165.html).

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.