Learn More
Gibco™ MEM, HEPES, GlutaMAX™ Supplement
Minimum Essential Medium is commonly used of all cell culture media
$180.00 - $1,524.00
Specifications
| Cell Line | HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts |
|---|---|
| Cell Type | Primary Rat Astrocytes |
| Classification | Animal Origin-free |
| Concentration | 1 X |
| Form | Liquid |
Description
MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. Gibco MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow's MEM, MEM α, DMEM, and Temin's Modification.
- Available with Earle's salts for use in CO2 incubator, or with Hank's salts for use without CO2
- Gibco MEM with GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format
- Modified as follows: with GlutaMAX™, HEPES, phenol red
- Made with Earle's salts
- Contains no proteins, lipids, or growth factors
- Requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
- Uses sodium bicarbonate buffer system (2.2g/L) and therefore requires 5-10% CO2 environment to maintain physiological pH
- Sustainable packaging
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room Temperature
Specifications
| HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts | |
| Animal Origin-free | |
| Liquid | |
| Standard Serum Supplementation | |
| Low Glucose, GlutaMAX, HEPES, Phenol Red | |
| Gibco, GlutaMAX | |
| Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
| Primary Rat Astrocytes | |
| 1 X | |
| MEM (Minimum Essential Medium) | |
| Sterile-filtered | |
| No Sodium Pyruvate | |
| Room Temperature |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.