missing translation for 'onlineSavingsMsg'
Learn More

Melting Point Standard, 47-49°C, MilliporeSigma™ Supelco™

Catalog No. p-7193371
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020667 1 g
111019503 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020667 Supplier Merck Emd Millipore Supplier No. 502961G
Only null left

Specifications

CAS 119-61-9
Vapor Pressure 1 mmHg (108°C)
Physical Form Neat
Linear Formula (C6H5)2 CO
MDL Number MFCD00003076
Melting Point 47°C to 49°C (±0.3 C); 47°C to 51°C
Boiling Point 305°C
Synonym Benzophenone; Diphenyl ketone; Diphenylmethanone; NSC 8077
Quantity 1 g
Formula Weight 182.22
Grade Analytical Standard
Show More Show Less