missing translation for 'onlineSavingsMsg'
Learn More

Melting point standard (121-123°C), MilliporeSigma™ Supelco™

Catalog No. 111020416
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020416 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020416 Supplier Merck Emd Millipore Supplier No. 761705G
Only null left

Melting point standard 121-123C can be used as a calibration standard while defining the purity of pharmaceutical chemicals.

Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care

Specifications

Ignition Point 1061°F
CAS 65-85-0
Vapor Pressure 10 mmHg (132°C)
Vapor Density 4.21 (vs. air)
Flash Point 121°C - Closed Cup
Physical Form Neat
Linear Formula C6H5 COOH
MDL Number MFCD00002398
Melting Point 121°C to 123°C (±0.3); 121°C to 125°C
Boiling Point 249°C
Synonym Benzoic acid
Type Analytical Standard
Quantity 5 g
Formula Weight 122.12
Grade Analytical Standard
Show More Show Less