Learn More
Meloxicam
CAS: 71125-38-7 | C14H13N3O4S2 | 351.40 g/mol
Supplier: Thermo Scientific Chemicals J6063514
Description
Meloxicam is developed for the treatment of rheumatoid arthritis and osteoarthritis. It is currently for the treatment of osteoarthritis and rheumatoid arthritis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water (22 mg/ml), DMSO (25 mg/ml), DMF, acetone(slightly soluble), ethanol(slightly soluble), and methanol(slightly soluble).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications
| Meloxicam | |
| 255°C | |
| C14H13N3O4S2 | |
| 25 g | |
| 14,5826 | |
| Soluble in water (22mg/ml),DMSO (25mg/ml),DMF,acetone(slightly soluble),ethanol(slightly soluble),and methanol(slightly soluble). | |
| CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O | |
| 351.40 | |
| CHEBI:6741 | |
| Powder |
| 71125-38-7 | |
| Yellow | |
| MFCD00868752 | |
| UN2811 | |
| meloxicam, mobic, metacam, movalis, meloxicamum, mobicox, mobec, meloxicam mobic, movatec, parocin | |
| DWMREKMVXIFPFM-ACCUITESSA-N | |
| (3E)-3-{hydroxy[(5-methyl-1,3-thiazol-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λâ¶,2-benzothiazine-1,1,4-trione | |
| 54677470 | |
| 351.39 |
Chemical Identifiers
| 71125-38-7 | |
| 351.40 | |
| DWMREKMVXIFPFM-ACCUITESSA-N | |
| 54677470 | |
| (3E)-3-{hydroxy[(5-methyl-1,3-thiazol-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λâ¶,2-benzothiazine-1,1,4-trione |
| C14H13N3O4S2 | |
| MFCD00868752 | |
| meloxicam, mobic, metacam, movalis, meloxicamum, mobicox, mobec, meloxicam mobic, movatec, parocin | |
| CHEBI:6741 | |
| CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber : DL0702000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only