missing translation for 'onlineSavingsMsg'
Learn More

Meloxicam

Catalog No. p-7041893
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6063506 5 g
AAJ6063514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6063506 Supplier Thermo Scientific Chemicals Supplier No. J6063506
Only null left

CAS: 71125-38-7 | C14H13N3O4S2 | 351.40 g/mol

Meloxicam is developed for the treatment of rheumatoid arthritis and osteoarthritis. It is currently for the treatment of osteoarthritis and rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (22 mg/ml), DMSO (25 mg/ml), DMF, acetone(slightly soluble), ethanol(slightly soluble), and methanol(slightly soluble).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.

Chemical Identifiers

CAS 71125-38-7
Molecular Formula C14H13N3O4S2
Molecular Weight (g/mol) 351.40
MDL Number MFCD00868752
InChI Key DWMREKMVXIFPFM-ACCUITESSA-N
Synonym meloxicam, mobic, metacam, movalis, meloxicamum, mobicox, mobec, meloxicam mobic, movatec, parocin
PubChem CID 54677470
ChEBI CHEBI:6741
IUPAC Name (3E)-3-{hydroxy[(5-methyl-1,3-thiazol-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λ⁶,2-benzothiazine-1,1,4-trione
SMILES CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O

Specifications

Melting Point 255°C
Color Yellow
Quantity 5 g
UN Number UN2811
Merck Index 14,5826
Solubility Information Soluble in water (22mg/ml),DMSO (25mg/ml),DMF,acetone(slightly soluble),ethanol(slightly soluble),and methanol(slightly soluble).
Formula Weight 351.39
Physical Form Powder
Chemical Name or Material Meloxicam
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber DL0702000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only