missing translation for 'onlineSavingsMsg'
Learn More

Melatonin, Powder, 96%, Spectrum™ Chemical

Catalog No. 18611519
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611519 1 g Amber Glass Bottle
18611523 5 g Amber Glass Bottle
18611522 25 g Amber Glass Bottle
18611521 100 g Amber Glass Bottle
18611524 500 g Amber Glass Bottle
18611520 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611519 Supplier Spectrum Chemical Mfg Cor Supplier No. M20951GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 73-31-4

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 g
Molecular Weight (g/mol) 232.27
Percent Purity 0.96
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Melatonin Powder

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.