missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Melanocyte-Stimulating Hormone-Release Inhibiting Factor, synthetic

Catalog No. AAJ60045MD
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60045MD 250mg
AAJ6004503 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60045MD Supplier Thermo Scientific Chemicals Supplier No. J60045MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibits α-melanocyte-stimulating hormone (α-MSH) release. Increases brain dopamine turnover and inhibits physiological and behavioral opioid effects in vivo. Role in vascular homeostasis. | 284.36 g/mol

Inhibits α-melanocyte-stimulating hormone (α-MSH) release. Increases brain dopamine turnover and inhibits physiological and behavioral opioid effects in vivo. Role in vascular homeostasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Molecular Weight (g/mol) 284.36
Quantity 250mg
Packaging Glass bottle
Form Lyophilized

RUO – Research Use Only