missing translation for 'onlineSavingsMsg'
Learn More

Melanin, MP Biomedicals™

Catalog No. p-4405117
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15534380 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15534380 Supplier MP Biomedicals Inc Supplier No. 0215534380

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8049-97-6
Molecular Formula C18H10N2O4
Molecular Weight (g/mol) 318.288
InChI Key XUMBMVFBXHLACL-UHFFFAOYSA-N
Synonym melanin, phaeomelanins, d0wq4g, melanin from sepia officinalis, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1; 2,5 .0; 13,17 .0; 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1^ 2,5 .0^ 13,17 .0^ 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone
PubChem CID 6325610
SMILES CC1=C2C3=C(C4=CNC5=C(C(=O)C(=O)C(=C45)C3=CN2)C)C(=O)C1=O

Specifications

Color Brown to Black
Quantity 100 mg
Formula Weight 1780
Physical Form Granular Powder
Chemical Name or Material Melanin