missing translation for 'onlineSavingsMsg'
Learn More

Melamine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012436
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012436 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012436 Supplier Merck Emd Millipore Supplier No. PHR12741G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Melamine is a nitrogen-rich chemical employed in the production of plastics, resins, laminates, adhesives as well as flame retardants. It may also be utilized as a fertilizer. It is one of the metabolites of cyromazine, which is approved for use as an insecticide on a wide range of vegetable crops.

Specifications

CAS 108-78-1
Flash Point 300°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C3H6N6
MDL Number MFCD00006055
Melting Point >300°C
Synonym 2,4,6-Triamino-1,3,5-triazine; sym-Triaminotriazine
Type Certified Reference Material
Quantity 1 g
Formula Weight 126.12
Grade Certified Reference Material
Show More Show Less