missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEKK4 Recombinant Rabbit Monoclonal Antibody (23GB4045)
GREENER_CHOICE

Catalog No. p-200056020
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552654 100 μL
MA552655 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552654 Supplier Invitrogen™ Supplier No. MA552654
Only null left

Rabbit Recombinant Monoclonal Antibody

MEKK4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

This gene encodes a TBP- (TATA box-binding protein) associated phosphoprotein that represses both basal and activated levels of transcription. The encoded protein is phosphorylated in vivo and this phosphorylation affects its interaction with TBP. This protein contains a histone fold motif at the amino terminus, a TBP-binding domain, and a glutamine- and alanine-rich region. The binding of DR1 repressor complexes to TBP-promoter complexes may establish a mechanism in which an altered DNA conformation, together with the formation of higher order complexes, inhibits the assembly of the preinitiation complex and controls the rate of RNA polymerase II transcription.

Specifications

Antigen MEKK4
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4045
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAP3K4
Gene Accession No. O08648, Q9Y6R4
Gene Alias D17Rp17; D17Rp17e; dJ473J16.1 (mitogen-activated protein kinase kinase kinase 4); JNKK1; KIAA0213; MAP three kinase 1; MAP/ERK kinase kinase 4; MAP2K4; Map3k4; MAPK/ERK kinase kinase 4; MAPKKK4; MEK kinase 4; Mek4b; MEKK 4; MEKK4; mitogen activated protein kinase kinase kinase 4; mitogen-activated protein kinase kinase kinase 4; mKIAA0213; MTK1; predicted protein of HQ0412; PRKMK4; PRO0412; RP17; Rp17a; SAPK/ERK kinase 1; SERK1; SSK2/SSK22 MAP kinase kinase kinase, yeast, homolog of; Tas; T-associated sex reversal
Gene Symbols MAP3K4
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAP3K4 (170-200AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26407, 308106, 4216
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less