missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Megestrol acetate

Supplier:  Thermo Scientific Chemicals 465790050

 View more versions of this product

Catalog No. AC465790050

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Megestrol acetate
210.0°C to 212.0°C
5 g
RQZAXGRLVPAYTJ-GQFGMJRRSA-N
(1R,3aS,3bR,9aR,9bS,11aS)-1-acetyl-5,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
384.51
595-33-5
C24H32O4
Solubility aq. soln.: .. Other solubilities: soluble in chloroform
CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(C)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)=O
384.52

Chemical Identifiers

595-33-5
RQZAXGRLVPAYTJ-GQFGMJRRSA-N
CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(C)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)=O
384.52
(1R,3aS,3bR,9aR,9bS,11aS)-1-acetyl-5,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Suspected of causing cancer.

May cause damage to organs through prolonged or repeated exposure.

GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 209-864-5

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only