Learn More
Thermo Scientific Chemicals Megestrol acetate
Supplier: Thermo Scientific Chemicals 465790010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Megestrol acetate | |
| 210.0°C to 212.0°C | |
| 1 g | |
| 384.51 |
| 595-33-5 | |
| C24H3204 | |
| Solubility aq. soln.: .. Other solubilities: soluble in chloroform |
Chemical Identifiers
| 595-33-5 |
Safety and Handling
GHS H Statement Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 209-864-5
RUO – Research Use Only