missing translation for 'onlineSavingsMsg'
Learn More

Mefenamic acid, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014903
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014903 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014903 Supplier Merck Emd Millipore Supplier No. 92574250MG
Pharmaceutical (small molecule)

Specifications

CAS 61-68-7
Physical Form Neat
Linear Formula C15H15 NO2
MDL Number MFCD00051721
Synonym 2-[(2,3-Dimethylphenyl)amino]benzoic acid; N-(2,3-Xylyl)anthranilic acid
Type Analytical Standard
Quantity 250 mg
Formula Weight 241.29
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard