missing translation for 'onlineSavingsMsg'
Learn More

Medisca Medroxyprogesterone Acetate, USP (Micro)

Catalog No. 14186095
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186095 100 g
14185992 5 g
14185595 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14186095 Supplier Medisca Supplier No. 142205*
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Medroxyprogesterone Acetate USP is a white to off-white, odorless, crystalline powder. Micronized so that 90% of particles are less than 10μm.

Medroxyprogesterone Acetate USP is a white to off-white, odorless, crystalline powder. Micronized so that 90% of particles are less than 10μm.

Specifications

Chemical Name or Material Medroxyprogesterone Acetate, USP (Micro)
Packaging Jar 500ml, White, HDPE, Round, 53-400, TRIVENI-DB, CT
CAS 71-58-9
Type Drug Standard
Molecular Weight (g/mol) 386.58
Quantity 100 g
Percent Purity 97 to 103 %
Grade USP