Learn More
Corning™ Medium 199 (Mod.)
Often used in vaccine production, virology, and for the culture of various non-transformed cells. It was originally formulated as the result of efforts to create a fully defined, animal origin-free medium.
$129.64
| Catalog Number | Mfr. No. | With Additives | Without Additives | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | With Additives | Without Additives | Price | Quantity & Availability | ||||
|
MT50050PB
|
Corning™
50050PB |
Glutamine, Earle's Balanced Salt Solution (EBSS) | No Sodium Bicarbonate |
Each for $129.64
|
|
||||
Description
Product manufactured under industry-leading standards for low endotoxin content and superior results. Medium 199 is often used in vaccine production, virology, and for the culture of various non-transformed cells. It was originally formulated as the result of efforts to create a fully defined, animal origin-free medium. Long-term use of this medium requires serum supplementation. This formulation contains Earle's salts and L-glutamine, and it does not contain sodium bicarbonate.Specifications
| 10000 mL | |
| 2° to 8°C |