Learn More
Corning™ Medium 199 (Mod.)
Catalog No. MT50050PB
Change view
Click to view available options
With Additives:
Glutamine, Earle's Balanced Salt Solution (EBSS)
Without Additives:
No Sodium Bicarbonate
Catalog No. With Additives Without Additives
MT50050PB Glutamine, Earle's Balanced Salt Solution (EBSS) No Sodium Bicarbonate
1 options
Catalog No. MT50050PB Supplier Corning™ Supplier No. 50050PB
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Often used in vaccine production, virology, and for the culture of various non-transformed cells. It was originally formulated as the result of efforts to create a fully defined, animal origin-free medium.
Product manufactured under industry-leading standards for low endotoxin content and superior results. Medium 199 is often used in vaccine production, virology, and for the culture of various non-transformed cells.  It was originally formulated as the result of efforts to create a fully defined, animal origin-free medium. Long-term use of this medium requires serum supplementation. This formulation contains Earle's salts and L-glutamine, and it does not contain sodium bicarbonate.
Specifications
With Additives Glutamine, Earle's Balanced Salt Solution (EBSS)
Without Additives No Sodium Bicarbonate
Quantity 10000 mL
Content And Storage 2° to 8°C